Learn More
Thermo Scientific™ Pierce™ Negative Ion Calibration Solution
Calibrate LTQ mass spectrometer instruments using a mixture of highly purified ionizable molecules designed for negative mode calibration.
Supplier: Thermo Scientific™ LSG88324
Description
Thermo Scientific™ Pierce ESI Negative Ion Calibration Solution is a mixture of highly purified ionizable molecules designed for negative mode calibration of Thermo Scientific™ LTQ series mass spectrometer instruments.
The Pierce ESI Negative Ion Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific Mass Spectrometers. The ESI Negative Ion Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.Highlights:
Strong peaks – mixture of SDS, sodium taurocholate and Ultramark 1621 in a acetonitrile/methanol/acetic solution
Ready-to-use – load the mass reference standard into a syringe and inject into the instrument
High-purity – mass spectrometry grade reagents in a non-leachable container
Stable – store at room temperature for more than one year
Specifications
Mass Spectrometer | |
Pierce™ | |
Peptides | |
Mass Spectrometry | |
Liquid |
Calibration Solution | |
Store between 2°C and 8°C. | |
Peptides | |
Calibration |
For Research Use Only. Not for use in diagnostic procedures.