Thermo Scientific™ Pierce™ Detergent Removal Spin Columns and Plates
Rapidly eliminate high concentrations of detergents with minimal sample loss with our spin column or filter-plate formats for centrifuge processing.
$177.00 - $834.00
Products 6
Catalog Number Mfr. No. Sufficient For Price Quantity  
Catalog Number Mfr. No. Sufficient For Price Quantity  
PI87776 View Documents Thermo Scientific™
87776
10 to 25uL sample with each column Each for $266.00
PI87777 View Documents Thermo Scientific™
87777*
25 to 100uL sample with each column Each for $333.00
PI87778 View Documents Thermo Scientific™
87778*
150 to 500uL sample with each column Each for $177.00
PI87779 View Documents Thermo Scientific™
87779*
500 to 1000uL sample with each column Each for $239.00
PI88304 View Documents Thermo Scientific™
88304*
192 samples of 25 to 100uL each Each for $834.00
PI87780 View Documents Thermo Scientific™
PI87780
1 to 2.5mL of typical detergent-containing sample Each for $510.00
Description
Description
Thermo Scientific™ Pierce Detergent Removal Resin efficiently binds to and removes high concentrations of detergents from 10μL to 1mL samples with minimal sample loss and is available prepacked into spin columns and 96-well spin filter plates.
The Pierce Detergent Removal Resin specifically binds a wide variety of detergents and surfactants that are commonly used in protein extraction and biological sample preparation and provide an especially convenient and rapid format for treating protein and peptide solutions to remove interfering detergents before downstream analysis by mass spectrometry and other techniques.
Highlights:
  • Fast and efficient protocol
  • Removes >95% of detergents with high protein recovery
  • Helps improves mass spectrometry peptide coverage
  • Best for processing samples with proteins or peptides at greater than 100μg/mL (for low-concentration samples, consider the Thermo Scientific HiPPR Detergent Removal Products)