missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Photo Evidence Scale

Perfect for precise measurement when photographing evidence.

Manufacturer:  Fisher Scientific PPS400

Catalog No. S99182



Description

Description

  • Durable 6 in. (15.2cm) vinyl rule
  • Scales are 0.02 in. thick, and calibrated in both inches and millimeters with crosshairs at each end
  • Scales feature a flat finish that eliminates reflections

Specifications

Specifications

6 in.
0.02 in.
0.05cm
White
Documents