missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenolphthalein (Certified ACS), Fisher Chemical™
$335.83 - $667.00
Chemical Identifiers
CAS | 77-09-8 |
---|---|
Molecular Formula | C20H14O4 |
Molecular Weight (g/mol) | 318.33 |
MDL Number | MFCD00005913 |
InChI Key | KJFMBFZCATUALV-UHFFFAOYSA-N |
Synonym | 3, 3-Bis(p-hydroxyphenyl)-1(3H)-isobenzofuranone, 3, 3-Bis(p-hydroxyphenyl) Phthallide |
PubChem CID | 4764 |
ChEBI | CHEBI:34914 |
IUPAC Name | 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one |
SMILES | OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
P79-100
|
Fisher Chemical
FLP79100 |
100 g | Glass Bottle |
Each for $335.83
Case of 6 Each for $2,014.98
|
|
||||
P79-500
|
Fisher Chemical
FLP79500 |
500 g | Poly Bottle |
Each for $667.00
Case of 6 Each for $4,002.00
|
|
||||
Chemical Identifiers
77-09-8 | |
318.33 | |
KJFMBFZCATUALV-UHFFFAOYSA-N | |
4764 | |
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one |
C20H14O4 | |
MFCD00005913 | |
3, 3-Bis(p-hydroxyphenyl)-1(3H)-isobenzofuranone, 3, 3-Bis(p-hydroxyphenyl) Phthallide | |
CHEBI:34914 | |
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
Specifications
258°C | |
Pass Test | |
C6H4COOC(C6H4-4-OH)2 | |
3, 3-Bis(p-hydroxyphenyl)-1(3H)-isobenzofuranone, 3, 3-Bis(p-hydroxyphenyl) Phthallide | |
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
318.33 | |
CHEBI:34914 | |
Certified ACS | |
Vapor Pressure: Negligible | |
White | |
Solid |
77-09-8 | |
C20H14O4 | |
MFCD00005913 | |
KJFMBFZCATUALV-UHFFFAOYSA-N | |
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one | |
4764 | |
318.33 | |
Glass Bottle | |
Pass Test | |
100 g | |
Phenolphthalein |