Promega™ pF9A CMV hRluc-neo Flexi™ Vector DFS Item
Designed specifically for high-level expression of proteins in mammalian cells from the CMV promoter with or without a selectable marker. The Flexi™ Vector System is a simple, directional cloning method for protein-coding sequences.
Manufacturer:  Promega™ C9361
Catalog No. PRC9361
Description
Description
  • You can choose between a variety of initial applications (e.g., bacterial protein, mammalian, or cell-free protein expression) and then transfer to others as required
  • Efficient transfer allows for direct use of recombinant clones, minimizing time wasted screening background colonies
  • Adaptable to high-throughput formats for large screening projects
Specifications
Specifications
20μg
−20°C