missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Petroleum Ether, B&J Brand™, for GC and pesticide residue analysis, >99.0% (C5 & C6 Isomers), Honeywell™ Burdick & Jackson
$988.30 - $1609.98
Chemical Identifiers
CAS | 8032-32-4 |
---|---|
Synonym | p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6001172
|
Honeywell-Burdick and Jackson
3171L |
6 x 1 L | Glass Bottle |
Case of 6 Each for $988.30
|
|
||||
6001171
|
Honeywell-Burdick and Jackson
3174 |
4 x 4 L | Glass Bottle |
Case of 4 Each for $1,609.98
|
|
||||
Chemical Identifiers
8032-32-4 | |
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Specifications
8032-32-4 | |
0.64 g/cm3 | |
6 x 1 L | |
Glass Bottle | |
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide | |
GC/Pesticide Residue Analysis |
100.0000% | |
35°C to 60°C | |
−18°C | |
UN1268 | |
>99% | |
Petroleum Ether, B&J Brand™, for GC and pesticide residue analysis, >99.0% (C5 & C6 Isomers) |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Petroleum distillates, n.o.s.