missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Argos Technologies PolarSafe™ PCR Cardboard Freezer Boxes

Designed to hold single PCR tubes or strips of 8.

Manufacturer:  Argos Technologies R3012A

 View more versions of this product

Catalog No. 10-987-067



Description

Description

  • The 1 in. height prevents the tubes from falling down inside the grids
  • Sized to fi t all standard stainless-steel and polycarbonate storage systems
  • Moisture repellant coating prevents water or liquid nitrogren from saturating box
  • Drain holes on bottom - allows for safe liquid nitrogen drainage and prevents excess moisuture build-up
  • Easy to write on surface for identification
  • Tight fitting lid-to-box fit
  • To maximize storage utility two of the boxes can be stacked inside each slot in the applicable stainless steel freezer racks
  • Can be used in mechanical freezers and liquid nitrogen tanks at temperatures to −196°C.
  • Cell opening: 8mm
Specifications

Specifications

14 x 14
5.25 in.
5.25 x 5.25 x 1 in.
Holding single PCR tubes or strips of 8
196 place PCR divider
1 in.
5.25 in.
PCR cardboard freezer box
Cardboard
196 tubes
Documents