Parker domnick hunter™ LC/MS Nitrogen Gas Generators
Specifically designed to meet the gas flow, purity, and pressure requirements of modern LC/MS instruments
Manufacturer:  Parker domnick hunter™ LCMS201W*
 View more versions of this product
Catalog No. 14-950-514
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Units use fully regenerative pressure swing adsorption (PSA) technology to trap oxygen, carbon dioxide, and moisture in a bed of carbon molecular sieve (CMS) that alternates between regeneration and purification modes.
  • Provide a continuous stream of clean, dry nitrogen without the need for secondary purification
  • Suitable for nebulizing, sheath, and drying gas
  • Digital interface; units are fully compatible with APCI and ESI interfaces
  • With FEP tubing for phthalate-free operation
  • Soundproofed compressor box
  • Space saving—can fit under a bench
  • LC/MS 15 is designed for use with Agilent™ 1100 Series LC/MS products
  • Some models include integral oil-free compressors–external supply is not needed
  • For those models without the integral compressor: 7barg specified outlet pressure, 9barg specified inlet pressure; 70L/min. minimum constant air requirement for 15 and 20 models, 130L/min. minimum constant air requirement for 30 and 40 models
  • International models also available
Compliance
CE, CSA and UL compliant and listed
Specifications
Specifications
Nitrogen Generator
32.5 in.
27.8 in.
50/60Hz
0.25 in. compression fitting
129kg
110V 50/60Hz
>99%
CE, CSA, UL
82.6cm
70.5cm
5° to 40°C
110V
20.1 in.
LCMS 20-1
101.5psi
Documents