missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Paraformaldehyde 90.0+%, TCI America™
$29.86 - $38.16
Chemical Identifiers
CAS | 30525-89-4 |
---|---|
Molecular Formula | CH2O |
Molecular Weight (g/mol) | 30.026 |
MDL Number | MFCD00133991 |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
Synonym | formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde |
PubChem CID | 712 |
ChEBI | CHEBI:16842 |
IUPAC Name | formaldehyde |
SMILES | C=O |
Chemical Identifiers
30525-89-4 | |
30.026 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00133991 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Specifications
Crystalline Powder | |
White | |
CH2O | |
2213 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
≥90.0% (T) |
30525-89-4 | |
25 g | |
MFCD00133991 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
C=O | |
30.026 | |
CHEBI:16842 | |
Paraformaldehyde |
Safety and Handling
EINECSNumber : (9)-1941
RTECSNumber : RV0540000
TSCA : Yes