Learn More
Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS
Paraformaldehyde, CH2O, CAS Number-30525-89-4, methylene oxide, paraformaldehyde, formalin, paraform, methyl aldehyde, formic aldehyde, formol, oxymethylene, methanal, oxomethane, 500mL, CHEBI:16842, H317-H318-H351
$158.70 - $282.85
Chemical Identifiers
CAS | 30525-89-4 |
---|---|
Molecular Formula | CH2O |
Molecular Weight (g/mol) | 30.026 |
MDL Number | MFCD00133991 |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
Synonym | formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde |
PubChem CID | 712 |
ChEBI | CHEBI:16842 |
IUPAC Name | formaldehyde |
SMILES | C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ61899AK
|
Thermo Scientific Chemicals
J61899AK |
250 mL |
Each for $158.70
|
|
|||||
AAJ61899AP
|
Thermo Scientific Chemicals
J61899AP |
500 mL |
Each for $282.85
|
|
|||||
Description
Paraformaldehyde, 4% in PBS is used to preserve tissues for routine histology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsParaformaldehyde, 4% in PBS is used to preserve tissues for routine histology.
Solubility
Miscible with water.
Notes
Store in a cool place. Incompatible with brass, steel, copper, acid anhydrides, strong oxidizing agents and strong reducing agents.
Chemical Identifiers
30525-89-4 | |
30.026 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00133991 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Specifications
Liquid | |
250 mL | |
MFCD00133991 | |
Miscible with water. | |
C=O | |
30.026 | |
CHEBI:16842 |
30525-89-4 | |
CH2O | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
Paraformaldehyde |
Safety and Handling
Causes serious eye damage, Suspected of causing cancer, May cause an allergic skin reaction
Serious eye damage/eye irritation (category 1), Carcinogenicity (category 2), Skin sensitisation (category 1)
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only