Parafilm M™ Wrapping Film
Waterproof, moistureproof film effectively seals labware.
$40.60 - $49.25
Specifications
Applications Effectively seals labware
Product Type Wrapping Film
Material Thermoplastic
View More Specs
Products 2
Catalog Number Mfr. No. Length (English) Width (English) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Length (English) Width (English) Width (Metric) Price Quantity  
S37440 Accessories View Documents Fisher Scientific
PM996
125 ft. 4 in. 10cm Each for $49.25
Case of 12 Each for $590.95
S37441 View Documents Fisher Scientific
ACAPM992CS24
250 ft. 2 in. 5cm Each for $40.60
Case of 24 Each for $974.80
Description
Description
  • Can be folded and twisted around sharp surfaces without rupturing
  • Becomes softer and more flexible as temperature increases
Specifications
Specifications
Effectively seals labware
Wrapping Film
Thermoplastic