missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Palladium hydroxide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03949601
Description
Acts as a catalyst for arylation reactions. It is employed in hydrogenolysis of benzyl-nitrogen bonds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Palladium hydroxide | |
12135-22-7 | |
0.25 g | |
H2O2Pd | |
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon | |
NXJCBFBQEVOTOW-UHFFFAOYSA-L | |
palladium(2+);dihydroxide | |
9942167 | |
99% |
Black | |
Powder | |
99% | |
MFCD00064599 | |
Insoluble in water. | |
[OH-].[OH-].[Pd++] | |
140.43 | |
140.42 | |
Odorless |
Chemical Identifiers
12135-22-7 | |
140.43 | |
NXJCBFBQEVOTOW-UHFFFAOYSA-L | |
9942167 |
H2O2Pd | |
MFCD00064599 | |
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon | |
[OH-].[OH-].[Pd++] |
Safety and Handling
EINECSNumber : 235-219-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only