missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Naphtholbenzein Indicator Solution, Fisher Chemical
Supplier: Fisher Chemical SN1500
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
p-Naphtholbenzein Indicator Solution | |
C27H18O2 | |
VDDWRTZCUJCDJM-PNHLSOANSA-N | |
(4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one | |
5941340 | |
Pass Test | |
Brown/Red | |
Liquid |
145-50-6,108-88-3,67-63-0,7732-18-5 | |
MFCD00078492 | |
C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O | |
374.439 | |
Glass Bottle | |
0.83g/cm³ | |
500 mL |
Chemical Identifiers
145-50-6 | |
374.439 | |
VDDWRTZCUJCDJM-PNHLSOANSA-N | |
(4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one |
C27H18O2 | |
MFCD00078492 | |
5941340 | |
C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O |