MilliporeSigma™ OxyBlot™ Protein Oxidation Detection Kit
Provides the reagents to perform the immunoblot detection of carbonyl groups
Supplier: MilliporeSigma™ S7150
Includes: Chemical and immunological reagents
Description
Provides the chemical and immunological reagents necessary to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidationSpecifications
Store all components at 2° to 8°C | |
Chemiluminescence | |
200 Assays |