Dynalon™ Oval Wash Bottle
Easy grip wash bottle
$18.35 - $55.00
Specifications
Includes | Integrally molded tube |
---|---|
Material | Polyethylene |
Autoclavable | Not Autoclavable |
Description
Molded polyethylene wash bottle with oval design for easy-grip action and single-handed operation.
- Discharge tube has fine stream diameter; only gentle pressure is required to eject and accurately direct the liquid
- Tube extends completely to the base of the bottle, so entire contents can be ejected
- Captured cap prevents drips and contamination
- Fill line on both sides of the bottle, so contents can be accurately gauged for repetitive work
- Withstands temperatures to 90°C
Specifications
Integrally molded tube | |
Polyethylene | |
Not Autoclavable |