Thermo Scientific™ Orion Star™ A326 pH/Dissolved Oxygen Portable Multiparameter Meter
Accurately measure pH, dissolved oxygen and temperature with our pH/DO/RDO portable meter for high performance in challenging field tests.
$3074.35 - $5463.46
Specifications
Outputs USB, RS232
AC Adapter Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
Accuracy (Barometer) ±6mmHg
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH) ±0.002
View More Specs
Products 2
Catalog Number Mfr. No. Description Includes Price Quantity  
Catalog Number Mfr. No. Description Includes Price Quantity  
13-645-566
View Documents Promotion Details
Thermo Scientific™
STARA3260
Orion Star A326 pH/RDO/DO portable meter Star A326 pH/RDO/DO meter, 4 AA batteries, literature CD, printed quick start guide, computer interface cable and meter test certificate
Each for $3,074.35
13-645-568
View Documents Promotion Details
Thermo Scientific™
STARA3265
Orion Star A326 pH/RDO/DO portable meter kit with ROSS Ultra Triode low maintenance gel pH/ATC electrode, RDO optical DO sensor, solutions, meter armor and field case Star A326 pH/RDO/DO Meter with Batteries; 8107UWMMD ROSS Gel-filled Epoxy pH/ATC Triode with 3 Meter Cable; 087010MD RDO Optical DO Probe with 3 Meter Cable; 910410 pH 4.01 Buffer Pouches; 910710 pH 7.00 Buffer Pouches; 911010 pH 10.01 Buffer Pouches; 810001 Orion ROSS electrode storage solution; 01100710 Orion 1413 μS conductivity standard pouches; 911110 rinse solution pouches; Protective armor with electrode holder; Hard carrying case; AA batteries (installed)
Each for $5,463.46
Description
Description
Get high accuracy and premium performance everywhere you need it with the Thermo Scientific™ Orion Star™ A326 pH/Dissolved Oxygen Portable Multiparameter Meter. This meter is designed for a wide range of pH, mV, ORP, dissolved oxygen and temperature testing and field applications. Evaluate samples in the most demanding locations using this waterproof meter with IP67-rated housing. Use the two meter channels to measure pH and dissolved oxygen simultaneously or view each channel separately. Log up to 5000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer.
Take your measurements at the source with our premium line of portable meters. The Thermo Scientific Orion Star A326 pH/RDO/DO Portable Meter offers an information-rich screen and user-friendly operation. This rugged, waterproof portable meter is ready to tackle the elements with you. Orion Star A326 pH/RDO/DO Portable Meters are designed to be easy-to-use, dependable field meters that offer advanced features and functions including a 5000 point date log with sample ID and user ID options, calibration and high/low limit alarms and password-protected measurement methods.
  • Combined convenience with the benefits of a pH meter and dissolved oxygen meter in one easy-to-use system
  • Measure pH, mV, relative mV or ORP with temperature on channel one and measure dissolved oxygen in percent saturation or mg/L with temperature using RDO optical DO probes or polarographic DO probes on channel two
  • Easily view individual measurements on a full screen display and quickly toggle to a split screen display to view multiple measurements simultaneously
  • Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values and calibration editing option that allows individual points to be fixed without a full recalibration
  • Calibrate dissolved oxygen using water-saturated air, air-saturated water, custom value (Winkler titration) and zero point options
  • Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 5,000 measurement sets with optional sample ID and user ID
  • Calibration log stores 10 most recent calibrations
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
USB, RS232
±6mmHg
±0.002
±0.1mg/L up to 8mg/L, ±0.2mg/L from 8 to 20mg/L, ±10% of reading up to 50mg/L
Yes, selectable
800 hr.
pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
1 point relative mV (RmV) mode, 1 point ORP mode
CE, TUV 3-1, FCC Class A
5000 with date and time stamp
LCD Graphic, Backlit
pH, mV, Relative mV, ORP with Temperature, Dissolved Oxygen (% Saturation), Dissolved Oxygen (mg/L with Temperature)
±2000.0 mV
0.1/0.01/0.001
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature)
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m
10 per channel with password protection
0.0 to 600.0 % saturation
0.1, 1 % saturation
±2 % saturation or ±2% of reading whichever is greater
4 AA batteries (included) or universal AC adapter (sold separately)
1mmHg
0.00 to 90.00mg/L
±2000.0 mV
0.1mV
0.01, 0.1mg/L
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
41°F to 113°F (ambient)
1 point temperature offset calibration
0.1°C, 0.1°F
3 Year Meter
1.2 lb.
Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
±0.2 mV or ±0.05% of reading whichever is greater
±0.2mg/L or ±2% of reading, whichever is greater
High/low limit alarm, calibration due alarm
400 to 850mmHg, automatic using built-in barometer or manual entry
4 AA (included)
Water-saturated air, air-saturated water, manual (Winkler titration), zero point
1 to 5
Transfer single, range or all data points to printer or computer
2
IP 67
0.0 to 600.0 % saturation
-2.000 to 20.000
-5.0°C to +105.0°C
7.000pH
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Non-volatile memory preserves data log, calibration log and meter settings
Orion Star A326 pH/mV/RmV/ORP/RDO Optical/Dissolved Oxygen/Temperature Portable Meter
±2 % saturation ≤ 200 % saturation, ±10 % saturation > 200 % saturation
0.0 to 500.0 % saturation
0.1, 1 % saturation
400 to 850mmHg
±2000.0 mV
0.00 to 50.00mg/L
5 to 85 %, non-condensing
0.01, 0.1mg/L
0.0 to 45.0 ppt, automatic correction using manual entry of sample salinity
±0.1°C
5°C to 45°C (ambient)
22.0°F to 221.0° F
Manual or automatic with ATC temperature probe
24cm x 10cm x 6.5cm (9.5 in. x 3.9 in. x 2.6 in.)
0.5 kg
SDS
missing translation for 'documents'
missing translation for 'documents'