missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ ISE Calibration Standards

Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.

$132.82 - $858.31

Specifications

Type Ion Selective Electrode Calibration Standard
Concentration 2ppm with TISAB
Packaging Bottle
For Use With (Equipment) Fluoride Ion Selective Electrodes
Description Fluoride Standard

Products 31 Description

Description

Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.

All standards are 1 pint (475mL) bottles, unless otherwise noted.

Warranty: Out of Box
Specifications

Specifications

Ion Selective Electrode Calibration Standard
Bottle
Fluoride Standard
2ppm with TISAB
Fluoride Ion Selective Electrodes

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.