Learn More
Thermo Scientific™ Orion™ Conductivity Standards and Solutions
Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
$201.46 - $1228.79
Specifications
Traceability to NIST | Yes |
---|---|
Type | Conductivity Standard |
Boiling Point | 100°C |
Solubility | Soluble |
Catalog Number | Mfr. No. | Packaging | Concentration | Conductivity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging | Concentration | Conductivity | Price | Quantity | |||
13-643-009
|
Thermo Scientific™
ORI011001 |
Bottle | — | — |
Pack of 5 Each for $201.46
|
|
|||
13-301-78
|
Thermo Scientific™
ORI011008 |
Bottle | 47 ppm as NaCl | 100 μS/cm |
Each for $278.83
|
|
|||
13-301-77
|
Thermo Scientific™
011007 |
Bottle | 692 ppm as NaCl | 1413 μS/cm |
Each for $278.83
|
|
|||
09-357-10
|
Thermo Scientific™
011006 |
Bottle | 7230 ppm as NaCl | 12.9 mS/cm |
Case of 5 Each for $278.83
|
|
|||
13-301-135
|
Thermo Scientific™
990106 |
Bottle | 0.1M KCl | 12.9 mS |
Each for $265.43
|
|
|||
13-303-150
|
Thermo Scientific™
ORI011005 |
Pouch | — | 111.9 mS/cm |
Each for $1,228.79
|
|
|||
Description
Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.
Conductivity Standards
- Traceable to NIST standard reference materials
- Designed to meet all end user requirements, including laboratory grade and DIN standards
- Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
- For use with Thermo Scientific™ Orion™ Conductivity Probes
Warranty: Out of Box
Specifications
Yes | |
Conductivity Standard | |
100°C | |
Soluble |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.