missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ AquaMate 8000 UV-Vis Spectrophotometer

Equip your lab with the accurate, easy to use Thermo Scientific™ Orion™ AquaMate 8000 UV-Vis Spectrophotometer to ensure the ideal instrument for water analysis and beyond

Manufacturer:  Thermo Scientific™ ORIAQ8000

Catalog No. 13-650-700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Equip your lab with the accurate, easy to use Thermo Scientific™ Orion™ AquaMate 8000 UV-Vis Spectrophotometer to ensure you have the ideal instrument for water analysis and beyond. This spectrophotometer offers 260 preprogrammed methods for ion concentration from absorbance plus wavelength scanning function and standard curve function for creating custom method using so virtually any brand of colorimetric reagent can be used. Utilize 190-1100 nm selectable wavelengths, 1.8 nm spectral bandwidth and vial flexibility with the included 3-position carousel for holding round and rectangular vials.

Pre-programmed Methods:

  • More than 260 pre-programmed methods offer easy and convenient measurements using Orion™ AQUAfast™, Merck™ or CHEMetrics™ chemistries
  • Pre-programmed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
  • Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries

Versatility:

  • Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50mm rectangular vials and an adjustable round vial holder that accommodates 13, 16, and 24mm round vials
  • Three USB connections provide the options to store methods and data to a USB memory stick, transfer data to a computer, and print hard copy data to an external printer
  • System is compatible with ink jet and laser printers with HP PCL 5 or greater

Verification:

  • Performance verification tests ensure wavelength accuracy and instrument functionality
  • Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP
  • Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards
  • Built-in test methods for stray light, noise and resolution are available to validate instrument performance further

Includes:

  • Methods on USB memory stick
  • Three-position cell holder
  • User documentation
  • Dust cover
  • USB cable
  • 110V, 220V and 240V power cords
Specifications

Specifications

±0.005A at 1.0A; <0.00025 at 0.0A
USB type A port for USB stick (front panel), USB type B port for computer (rear panel), USB type A port for printer (rear panel)
Dual silicon photodiodes
<0.0005A/hr
9.8 in.
50/60Hz
AquaMate 8000 UV-Vis Spectrophotometer
Xenon flash lamp (5 years)
<0.00050 at 1.0 A; <0.00080 at 2.0 A RMS at 260nm
Up to 3.5A at 260nm
1.8nm
100 to 240V
190 to 1,100nm
10 to 4,200nm/min
19 lb.
11.8 in.
Compatible with ink jet and laser printers with HP PCL 5 or greater
AquaMate 8000 UV-Vis Spectrophotometer
Graphical with LCD backlight—9.7 x 7.1cm (3.8 x 2.8 in.)
100 to 240V 50/60Hz
25cm
Methods on USB memory stick; 6-position cell holder; 3-position cell holder; 1-position cell holder; User documentation; Dust cover; USB cable; 110V and 220V power cords
Sealed membrane with tactile response keys
0.2; 0.5; 1.0; 2.0; 3.0; 5.0nm
Dual beam—internal reference detector
-0.5 to 5.0 A ; -1.5 to 125 %T; ±9999 C
<0.08 %T at 220nm and 340nm; <0.1 %T at 340nm and 400nm; <1.0 %T at 198nm
±1.0nm
±0.5nm
11,000nm/min
8.6kg
30cm
Documents