OPTIZYME™ 10X Buffer 3, Fisher BioReagents™
Supplier: Thermo Fisher Scientific BP80861
Description
1X Buffer Composition:
50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl, 0.1mg/mL BSA
Specifications
50mM Tris HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl, 0.1mg/mL BSA | |
5 x 1 mL | |
Liquid |
-20°C | |
Buffer 3 |