Learn More
Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles
Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles.
$874.06 - $4654.72
Specifications
Additive | Proprietary anionic surfactant |
---|---|
Certifications/Compliance | Includes Certificate of Analysis |
Content And Storage | Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
For Use With (Application) | Covalent Coupling |
Catalog Number | Mfr. No. | Quantity | Diameter (Metric) | Surface Acid Loading | Concentration | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Diameter (Metric) | Surface Acid Loading | Concentration | Price | Quantity | ||
09981057
|
Thermo Scientific™
73003305100250 |
15 mL | 2 μm | High Acid | 10% |
Each for $874.06
|
|
||
09981058
|
Thermo Scientific™
73003305100350 |
100 mL | 2 μm | High Acid | 10% |
Each for $3,801.34
|
|
||
09981060
|
Thermo Scientific™
73003420100250 |
15 mL | 3 μm | High Acid | 10% |
Each for $925.78
|
|
||
09981061
|
Thermo Scientific™
73003420100350 |
100 mL | 3 μm | High Acid | 10% |
Each for $4,654.72
|
|
||
09981036
|
Thermo Scientific™
83000520100290 |
15 mL | 0.2 μm | High Acid | 10% |
Each for $978.53
|
|
||
09981037
|
Thermo Scientific™
83000520100390 |
100 mL | 0.2 μm | High Acid | 10% |
Each for $3,491.03
|
|
||
09981033
|
Thermo Scientific™
83000550100290 |
15 mL | 0.2 μm | Medium Acid | 10% |
Each for $925.78
|
|
||
09981034
|
Thermo Scientific™
83000550100390 |
100 mL | 0.2 μm | Medium Acid | 10% |
Each for $3,491.03
|
|
||
09981045
|
Thermo Scientific™
83000720100290 |
15 mL | 0.3 μm | High Acid | 10% |
Each for $925.78
|
|
||
09981046
|
Thermo Scientific™
83000720100390 |
100 mL | 0.3 μm | High Acid | 10% |
Each for $3,491.03
|
|
||
09981044
|
Thermo Scientific™
83000750100290 |
15 mL | 0.3 μm | Medium Acid | 10% |
Each for $978.53
|
|
||
09981043
|
Thermo Scientific™
83000750100390 |
100 mL | 0.3 μm | Medium Acid | 10% |
Each for $3,491.03
|
|
||
09981054
|
Thermo Scientific™
83000920100290 |
15 mL | 0.4 μm | High Acid | 10% |
Each for $925.78
|
|
||
09981055
|
Thermo Scientific™
83000920100390 |
100 mL | 0.4 μm | High Acid | 10% |
Each for $3,491.03
|
|
||
09981048
|
Thermo Scientific™
83000970100290 |
15 mL | 0.4 μm | Low Acid | 10% |
Each for $925.78
|
|
||
09981049
|
Thermo Scientific™
83000970100390 |
100 mL | 0.4 μm | Low Acid | 10% |
Each for $3,491.03
|
|
||
09-980-863
|
Thermo Scientific™
93000570100390 |
100 mL | 0.2 μm | Low Acid | 10% |
Each for $3,277.97
|
|
||
09980684
|
Thermo Scientific™
W050CA |
15 mL | 0.5 μm | Medium Acid | 4% |
Each for $925.78
|
|
||
09980685
|
Thermo Scientific™
W050CB |
100 mL | 0.5 μm | Medium Acid | 4% |
Each for $3,491.03
|
|
||
09980686
|
Thermo Scientific™
W080CA |
15 mL | 0.85 μm | Medium Acid | 4% |
Each for $925.78
|
|
||
09980687
|
Thermo Scientific™
W080CB |
100 mL | 0.85 μm | Medium Acid | 4% |
Each for $3,491.03
|
|
||
09980688
|
Thermo Scientific™
W090CA |
15 mL | 0.9 μm | High Acid | 4% |
Each for $925.78
|
|
||
09980689
|
Thermo Scientific™
W090CB |
100 mL | 0.9 μm | High Acid | 4% |
Each for $3,491.03
|
|
||
09980690
|
Thermo Scientific™
W400CA |
15 mL | 4.0 μm | High Acid | 4% |
Each for $925.78
|
|
||
09980691
|
Thermo Scientific™
W400CB |
100 mL | 4.0 μm | High Acid | 4% |
Each for $3,491.03
|
|
||
09980692
|
Thermo Scientific™
W500CA |
15 mL | 5.0 μm | High Acid | 4% |
Each for $925.78
|
|
||
09980693
|
Thermo Scientific™
W500CB |
100 mL | 5.0 μm | High Acid | 4% |
Each for $3,491.03
|
|
||
Description
Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles. Particles contain carboxylic acid groups for covalent coupling and can be used directly from the bottle in a variety of applications.
- Parking area combines surface area and acid content to give you the surface acid distribution that is useful for assay optimization
- Surface area per gram of a particle varies inversely with the particle diameter
- Acid content refers to the titrated milliequivalents per gram (mEq/g) and is weight-based
- For most applications, particles can be used directly from the bottle without any pre-cleaning
- Proprietary anionic surfactant does not interfere with binding of proteins nor cause proteins to desorb from particle surfaces or cause proteins to desorb from particle surfaces
- Various surface acid concentrations available to help optimize reagent development efforts
- Particles have been used in and are equally successful in molecular biology and research applications
Parking Area Properties
- Low Acid Content—60–100 Parking Area—Hydrophobic surface, more colloidally stable than plain polystyrene
- Medium Acid Content—35–59 Parking Area—Hydrophobic and hydrophilic areas; good colloidal stability and good covalent coupling
- High Acid Content—10–34 Parking Area—Hydrophilic surface; excellent colloidal stability; excellent covalent coupling
Recommended for:
- Clinical diagnostics
- Molecular biology
- Nephelometric, turbidimetric and slide agglutination assays
Note:
When using Opti-Link particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.
Specifications
Proprietary anionic surfactant | |
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. | |
Covalent Coupling | |
Specialty Particles | |
1.05g/cm³ |
Includes Certificate of Analysis | |
Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. | |
Polystyrene Sulfate | |
Package Insert Sheet containing Certicate of Analysis | |
Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |