missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Starter pH Pen Meters

Convenient pH testing. OHAUS Starter pH Pen Meters offer a convenient way to measure pH, or the relative acidity or alkalinity of a sample at a given temperature. The pH pen meters can be completely operated with just one hand, freeing up your other hand to handle samples.

Supplier:  OHAUS™ 30073970

 View more versions of this product

Catalog No. S05084



Description

Description

  • Fast, straightforward results
  • Durable IP67 waterproof design
  • Equipped with wrist strap for security
  • Automatic Shutdown after six minutes of non-use
  • Requires four AG13 1.5V Micro Alkaline Batteries
  • Size/Weight: 185 × 45 × 38mm, 110g
  • Includes buffer powder pack (ST20)
Specifications

Specifications

±0.1
0 to 14
0.0°C to 99.0°C
110 g
Yes
Portable pH Meter
Measuring pH, or the relative acidity or alkalinity of asample at a given temperature
0.1
Dual
0.24 lb.
4 x AG13 1.5V Micro Alkaline
SDS
Documents

Documents

Promotions

Promotions