Please login to your online account to display your discounted pricing
OHAUS™ Scout™ STX Portable Balances with Touchscreen Display
The all new Ohaus Scout with intuitive color touchscreen interface offers superior overload protection in a sleek, low profile and stackable design.
$649.90 - $1,028.70
Specifications
Material ABS housing, stainless steel pan
Connection Type Bluetooth, Ethernet, USB Host, USB Device, RS234
Display Type Touch Screen
View More Specs
Products
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity    
S14514
ohaus™
30253014
STX2201 1 sec. 2200g 0.1g Each for $649.90
S14515
ohaus™
30253015
STX6201 1 sec. 6200g 0.1g Each for $1,028.70
S14507
ohaus™
30253007
STX222 1 sec. 220g 0.01g Each for $727.65
Description & Specifications
Specifications
Material ABS housing, stainless steel pan
Connection Type Bluetooth, Ethernet, USB Host, USB Device, RS234
Display Type Touch Screen
  • Fast response time and repeatable weighing results
  • Rapid stabilization and high resolution
  • Five major connectivity options
  • Additional software application modes and added weighing units complement the balance and set a new standard in portable weighing technology
  • Available with or without draftshield
  • Stackable design
  • Leveling bubble with adjustable feet
  • Kesnington security slot cable sold separately
  • Calibration mass included(all models up to 620g)
  • 12 different weighing units:g,mg,kg,lb:oz,N,ct,Grn, dwt, ozt, custom unit
  • Powered by AC adapter (included) or 4 AA batteries (not included)
  • Integrated weigh-below hook
  • Transportation/storage lock
Applications
Weighing, density determination, molar mass