missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ CS Series Compact Scales

Lightweight, portable electronic scales, ideal for work outside of the lab

$214.89 - $468.40

Specifications

Type CS Compact Scale
Stabilization Time 3 sec.
View More Specs

Includes: Three AA batteries, Stainless-steel weighing pan, AC adapter

Products 5
Catalog Number Mfr. No. Capacity Includes Model Readability Price Quantity  
Catalog Number Mfr. No. Capacity Includes Model Readability Price Quantity  
01-918-019 View Documents OHAUS™
80500633
200 g Postal charts, AC adapter, three AA batteries, stainless-steel weighing pan CS-200P 0.1g Each for $432.83
01918021 View Documents OHAUS™
80500635
11 lb., 5 kg Postal charts, AC adapter, three AA batteries, stainless-steel weighing pan CS5000P 1g Each for $275.70
01-919-34 View Documents OHAUS™
72212664
2 kg (72 oz.) AC adapter, three AA batteries, stainless-steel weighing pan CS2000 1g Each for $402.46
01-919-33 View Documents OHAUS™
72212663
200 g AC adapter, three AA batteries, stainless-steel weighing pan CS200-001 0.1g Each for $214.89
01-919-35 View Documents OHAUS™
72212665*
5 kg, 11.02 lb. AC adapter, three AA batteries, stainless-steel weighing pan CS5000-001 1g Each for $468.40
Description

Description

Lightweight, portable electronic scales, ideal for work outside of the lab

Suited for a wide range of applications, including quality control, formulations, postal weighing, forensic testing and soil and water sampling, and dietary needs.

  • Easy to operate with simple two-button operation with standard stainless steel weighing pan
  • Three low-profile models with weighing capacities from 200 to 5000g Low-battery indicator, overload/underload indicators, tactile feedback keyboard and auto shutoff Quick, easy calibration allows user to adjust for changes in location and temperature
  • Rugged construction provides overload protection up to 150% of capacity Powered by three AA batteries or AC adapter (both included)
  • Weighs in g, lb:oz, Newton, oz
  • Net weight: 0.91 lb. (0.47kg)
  • Dimensions (L × W × H): 7.625 × 5.312 × 1.5 in. (19.3 × 13.5 × 3.9cm)
  • Manufactured in ISO 9001 registered facility

Service Plan

Service agreement available.

Certifications

ISO 9001 compliant

Warranty

One-year (U.S. and Canada)

Specifications

Specifications

CS Compact Scale
3 sec.