missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oakton™ pHTestr 50 Waterproof Pocket pH Tester

Oakton pHTestr 50 has a replaceable double junction sensor for longer life, an expanded number of applications, and easy maintenance. Versatile, compact design works in the field or laboratory

Supplier:  Oakton™ 3563415

Catalog No. 11-747-327



Includes: Protective plastic storage case, lanyard, and 4 AAA batteries.

Description

Description

  • Redesigned leakproof cap functions as a sample cup, hydrating cup, and stabilizing base to stand the meter upright
  • Easy-to-navigate interface for quick setup and calibration
  • Replaceable, double-junction electrode sensors save time and money
  • Automatic temperature compensation ensures accurate reading in varying environments

Warranty

1 Year

Specifications

Specifications

Oakton pHTestr 50 Waterproof Pocket pH Tester, Premium 50 Series
±0.5°C, ±0.9°F for 0.0°C to 50.0°C
Water quality testing, buffer and reagent production, electrochemistry
-1 to 15
Up to 3
32°F to 140°F
Graphics, Dot Matrix 80 x 100 pixels
Hold Data
1 Year
Valox™
Integral electrode
Checkmark indicator
41°F to 113°F (ambient)
1
pH Meter
Four AAA alkaline batteries
±0.01
Protective plastic storage case, lanyard, and 4 AAA batteries.
pH, Temperature
0.01
0°C to 60°C
0.1°F/°C
Yes
IP67
Membrane
50 Series
English
Automatic
5°C to 45°C (ambient)
4 AAA alkaline
250+ hr.
SDS
Documents

Documents

Promotions

Promotions