missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
o-Xylene, CHROMASOLV™ Plus, for HPLC, 98%, Honeywell Riedel-de Haen
CHROMASOLV™ Plus, for HPLC, 98%
Supplier: Honeywell-Chromasolv 2958841L
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Specifications
o-Xylene | |
CHROMASOLV™ Plus, for HPLC, 98% | |
max. 0.01 at 350 nm, max. 0.05 at 325 nm, max. 0.40 at 300 nm, max. 1.00 at 288 nm | |
0.879Â g/mLÂ at 20Â °C(lit.) | |
1 L | |
Glass Bottle | |
C6H4(CH3)2 | |
UN1307 | |
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
CC1=CC=CC=C1C | |
106.17 | |
CHEBI:28063 | |
>97.5% |
-26°C to -23°C (literature) | |
95-47-6 | |
100.0000% | |
143°C to 145°C | |
30°C | |
C8H10 | |
MFCD00008519 | |
1815558 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
1,2-xylene | |
7237 | |
106.17 | |
HPLC |
Chemical Identifiers
95-47-6 | |
106.17 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
7237 | |
1,2-xylene |
C8H10 | |
MFCD00008519 | |
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
CHEBI:28063 | |
CC1=CC=CC=C1C |
Safety and Handling
ShelfLife : 720 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Xylenes
EINECSNumber : 202-422-2
Recommended Storage : Room Temp