missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-tert-Butyl-L-threonine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6352903
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
O-tert-Butyl-L-threonine methyl ester hydrochloride | |
C9H20ClNO3 | |
4716005 | |
SDHKEUUZUMQSAD-HHQFNNIRSA-N | |
methyl (2S,3R)-2-amino-3-[(2-methylpropan-2-yl)oxy]butanoate;hydrochloride | |
16218568 | |
95% | |
Solid |
71989-43-0 | |
MFCD00077111 | |
h-thr tbu-ome.hcl, h-thr tbu-ome hcl, 2s,3r-methyl 2-amino-3-tert-butoxy butanoate hydrochloride, o-tert-butyl-l-threonine methyl ester hydrochloride, o-t-butyl-l-threonine methyl ester hydrochloride, methyl o-1,1-dimethylethyl-l-threoninate hydrochloride, l-threonine, o-1,1-dimethylethyl-, methyl ester, hydrochloride, h-thr but-ome hcl, h-thr t-bu-ome*hcl;, ksc491o2b | |
Cl.COC(=O)[C@@H](N)[C@@H](C)OC(C)(C)C | |
225.71 | |
225.71 | |
1 g |
Chemical Identifiers
71989-43-0 | |
225.71 | |
SDHKEUUZUMQSAD-HHQFNNIRSA-N | |
16218568 | |
Cl.COC(=O)[C@@H](N)[C@@H](C)OC(C)(C)C |
C9H20ClNO3 | |
MFCD00077111 | |
h-thr tbu-ome.hcl, h-thr tbu-ome hcl, 2s,3r-methyl 2-amino-3-tert-butoxy butanoate hydrochloride, o-tert-butyl-l-threonine methyl ester hydrochloride, o-t-butyl-l-threonine methyl ester hydrochloride, methyl o-1,1-dimethylethyl-l-threoninate hydrochloride, l-threonine, o-1,1-dimethylethyl-, methyl ester, hydrochloride, h-thr but-ome hcl, h-thr t-bu-ome*hcl;, ksc491o2b | |
methyl (2S,3R)-2-amino-3-[(2-methylpropan-2-yl)oxy]butanoate;hydrochloride |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only