missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Phenylenediamine, Fisher BioReagents

Free Base

Manufacturer:  Fisher BioReagents BP253750

 View more versions of this product

Catalog No. BP253750




Description

Description

  • A chromogenic peroxidase substrate useful for ELISA and other enzyme immunoassays
  • Commonly used as a substrate for horseradish peroxidase (HRP) in ELISAs
  • When oxidized, OPD produces a color change that is detectable by spectrophotometry
  • Color intensity will increase over time as OPD is oxidzed by peroxidase
Specifications

Specifications

o-Phenylenediamine
256°C
100°C
95-54-5
C6H4(NH2)2
o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine
C1=CC=C(C(=C1)N)N
108.144
CHEBI:34043
Solid
Amber Glass
Cream
50g
C6H8N2
15, 7396
GEYOCULIXLDCMW-UHFFFAOYSA-N
benzene-1,2-diamine
7243
108.1

Chemical Identifiers

95-54-5
108.144
o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine
CHEBI:34043
C1=CC=C(C(=C1)N)N
C6H8N2
GEYOCULIXLDCMW-UHFFFAOYSA-N
7243
benzene-1,2-diamine
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 6.1, : Poison

Recommended Storage : RT

SDS Documents
Promotions

Promotions