Learn More
Thermo Scientific™ Nunc™ Cell Culture/Petri Dishes
Perform a variety of applications across a broad range of sizes and styles to suit a variety of experimental needs.
$607.21 - $3403.14
Specifications
Closure Included | Closure Included |
---|---|
Shape | Round |
Sterility | Sterile |
Type | Cell Culture Dish |
Includes | Lid |
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
12-565-90
|
Thermo Scientific™
150318 |
35x10mm Dish, Nunclon™ Delta |
Case of 500 Each for $926.60
|
|
|||||
12-565-91
|
Thermo Scientific™
153066 |
35x10mm Dish, Nunclon™ Delta |
Case of 500 Each for $961.88
|
|
|||||
12-565-92
|
Thermo Scientific™
174926 |
35x10mm Dish, Nunclon™ Delta, gridded |
Case of 500 Each for $1,194.47
|
|
|||||
12-565-93
|
Thermo Scientific™
171099 |
35x10mm Dish, Non-treated |
Case of 500 Each for $1,095.58
|
|
|||||
12-565-97
|
Thermo Scientific™
174888 |
60x15mm Dish, Cell culture-treated Permanox |
Case of 500 Each for $3,403.14
|
|
|||||
12-565-94
|
Thermo Scientific™
150326 |
60x15mm Dish, Nunclon™ Delta |
Case of 400 Each for $882.49
|
|
|||||
12-565-95
|
Thermo Scientific™
150288 |
60x15mm Dish, Nunclon™ Delta |
Case of 400 Each for $822.95
|
|
|||||
12-565-96
|
Thermo Scientific™
169558 |
60x15mm Dish, Nunclon™ Delta, gridded |
Case of 400 Each for $1,306.49
|
|
|||||
12-565-020
|
Thermo Scientific™
150350 |
100x15mm Dish, Nunclon™ Delta |
Case of 150 Each for $607.21
|
|
|||||
12-565-100
|
Thermo Scientific™
168381 |
150x21mm Dish, Nunclon™ Delta |
Case of 80 Each for $757.63
|
|
|||||
12-567-650
|
Thermo Scientific™
172931 |
100x21mm Dish, Nunclon™ Delta |
Case of 240 Each for $921.42
|
|
|||||
Description
With wide selections in size and style, Thermo Scientific™ Nunc™ Cell Culture Dishes suit a multitude of experimental needs in your cell culture routine. Perform a variety of applications in dishes with the Nunclon™ Delta surface treatment that promotes maximum adhesion for a broad range of cell types.
Features:
- Top and bottom stacking rings for easier handling and more stable stacking
- Sterile and non-pyrogenic
- Available with or without air vents for gas exchange
- 2 x 2mm grid for cloning experiments and determination of plating efficiency
Ordering Information:
174926 and 169558 feature 2x2mm grid
171099 for suspension cell culture - not Nunclon Delta treated
Includes:
A certificate that assures user of biological performance as well as traceability.
Warranty: 90 days
Specifications
Closure Included | |
Sterile | |
Lid |
Round | |
Cell Culture Dish |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.