missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nuclease-Free Water, Promega™
Quality tested and certified free of DNase and RNase activity.
$92.31 - $148.73
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Description
- Quality tested and certified free of DNase and RNase activity
Chemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Specifications
7732-18-5 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Water, Nuclease-Free |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
2 x 25 mL |
Safety and Handling
Recommended Storage : Below 30°C