missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ SYPRO™ Ruby Protein Blot Stain
Compatible with immunostaining (Western blotting), microsequencing and mass spectrometry
Supplier: Invitrogen™ S11791
Description
SYPRO™ Ruby protein blot stain is a highly sensitive, ready-to-use fluorescent stain for proteins on nitrocellulose or PVDF membranes. Stained proteins can be viewed with a standard UV transilluminator or with a laser scanner.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature and protect from light. | |
Fluorescent | |
Protein Blot Stain | |
Room Temperature |
In-Blot Detection | |
SYPRO Ruby | |
SYPRO™ | |
Protein |
For Research Use Only. Not for use in diagnostic procedures.