Invitrogen™ Novex™ PowerEase™ 300W Power Supply (115 VAC) DFS Item
Designed for high-throughput gel electrophoresis
Manufacturer:  Invitrogen™ LSPS0300
Catalog No. PS0300
Includes: Power supply and a quick reference card
Description
Description
The PowerEase 300W Power Supply is a fully programmable power supply with a straight-forward, intuitive interface that makes the powering of gel runs a simple and easy process.
  • Constant voltage, current, or power settings
  • Built-in timer for walk-away gel electrophoresis
  • Up to 10 custom programs with 10 steps each
  • Four sets of output jacks that are compatible with most electrophoresis devices
Easy to Use
  • Simply set a constant voltage, current, or power to begin a run
  • Easily accommodates the running and transferring of eight mini-gels, such as Bolt™ Bis-Tris Plus gels, or 16 midi-gels using the XCell4 SureLock™ Midi-Cell
Programming Capabilities
  • Accommodates up to 10 user-defined programs
  • Each program can include up to 10 steps, for precise control over electrophoresis conditions
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Specifications
Novex Semi-Dry Blotter, XCell II Blot Module, XCell SureLock Mini, XCell4 SureLock Midi, XCell6 MultiGel
Power supply and a quick reference card
300W
4
Documents