missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ NuPAGE™ MOPS SDS Running Buffer (20X) DFS Item

Made with high-purity reagents and are strictly quality controlled

$104.00 - $800.00

Specifications

Buffer Running Buffers
Concentration 20X
Formulation 50mM MOPS, 50mM Tris Base, 0.1% SDS, 1mM EDTA, pH 7.7
For Use With (Application) 1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Includes NuPAGE MOPS SDS Running Buffer (20X)
View More Specs

Includes: NuPAGE MOPS SDS Running Buffer (20X)

Requires: The buffers are provided as a concentrated solution and require simple dilution with deionized water before use.


Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NP0001 View Documents Invitrogen™
LSNP0001
500mL Each for $104.00
NP000102 View Documents Invitrogen™
LSNP000102
5L Each for $800.00
Description

Description

NuPAGE MOPS SDS Running Buffer (20X) is formulated for running proteins on NuPAGE Novex Bis-Tris gels only. NuPAGE™ MOPS SDS Running Buffer is recommended for separating small- to medium-sized proteins.

  • Get consistent, convenient, high-quality results
  • Pre-mixed NuPAGE buffers are convenient way to ensure high-quality, consistent electrophoresis results

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Running Buffers
50mM MOPS, 50mM Tris Base, 0.1% SDS, 1mM EDTA, pH 7.7
NuPAGE MOPS SDS Running Buffer (20X)
Store at 4° to 25°C
20X
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
pH 7.7

For Research Use Only. Not for use in diagnostic procedures.