missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ Gel Loading Tips DFS Item

For easy and accurate loading of samples

$119.00 - $214.00

Specifications


Includes: One pack (200/pack) of Standard, Round gel loading tips.

Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
LC1002 View Documents Invitrogen™
LSLC1002
Each for $214.00
LC1001 View Documents Invitrogen™
LSLC1001
Each for $119.00
Description

Description

Tips are RNase/DNase-free and non-autoclavable. Standard, round can be used for easy and accurate loading of samples onto 1.0mm and 1.mm gels. Loading volume is 0.5 to 200 μL.

Applications:

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Room Temperature

Specifications

Specifications

0.0
icon-pdf.png

For Research Use Only. Not for use in diagnostic procedures.