missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Dynabeads™ Protein A for Immunoprecipitation DFS Item

Dynabeads Protein A beads are uniform, 2.8-μm superparamagnetic beads with recombinant Protein A (∼45 kDa) covalently coupled to the surface.

Supplier:  Invitrogen™ 10008D

 View more versions of this product

Catalog No. 10008D



Description

Description

Dynabeads Protein A beads are uniform, 2.8-μm superparamagnetic beads with recombinant Protein A (∼45 kDa) covalently coupled to the surface. Dynabeads Protein A beads provide a great alternative to Sepharose or agarose resins for immunoprecipitation (IP) combined with your own antibody, and both manual and automated protocols are available.

Benefits of Dynabeads Protein A beads include:
Fast and easy—IP in less than 40 minutes with no columns, centrifugations, or time-consuming pre-clearing required
High performance—high target protein yield with low antibody consumption
Low background­—very low non-specific binding with a high signal-to-noise ratio
High reproducibility—uniform beads ensure the most consistent results
Automation ready—automation protocols available with the KingFisher Sample Purification instruments

Gentle separation causes minimal physical stress to proteins
The magnetic separation technology utilized by Dynabeads Protein A is rapid and gentle, causing minimal physical stress to target proteins. This permits the isolation and concentration of labile composites that might otherwise dissociate or be damaged by proteases during long incubation times. Native protein conformation and large protein complexes are preserved.

Binding strength and antibody binding capacity
Dynabeads Protein A beads allow for the isolation of most mammalian immunoglobulins (Ig). The amount of Ig captured depends on the concentration of Ig in the starting sample, and the type and source of the Ig. 100 μL of Dynabeads Protein A will isolate approximately 25–30 μg human IgG from a sample containing 20–200 μg IgG/mL. Predominantly Fc-binding allows optimal Ig orientation.

Dynabeads magnetic beads are not porous in nature such as Sepharose or agarose resins. Thus, the antibodies bind to the smooth outer surface of the beads and are available to bind to the target protein. This translates to a low antibody consumption while obtaining a high yield of the target protein. The smooth bead surface is also the reason for the low non-specific binding that Dynabeads magnetic beads are known for. The recombinant protein A on the beads does not contain albumin binding sites, thus albumin is not co-purified during the procedure.

Dynabeads immunoprecipitation protocol is fast and easy
The manual IP protocol is simple and can be performed in less than 40 minutes with 3 easy bind-incubate-elute steps. The procedure does not require any pre-clearing due to the low non-specific binding and uses very little antibody while still obtaining a high IP yield, thus saving both time and cost per IP.

The antibody-coated beads can be used in a variety of IP applications such as Co-IP, chromatin IP (ChIP/ChIP sequencing), RNA IP (RIP, RIP sequencing), small-scale IgG purification, and protein purification. The isolated pure target protein can be used in downstream western blot, mass spectrometry analysis, sequencing, etc.

Automation-ready for use with KingFisher purification systems
The 2.8-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher sample purification systems

Commercial supply
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads Protein A beads in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or read more on our Dynabeads OEM information page.

Learn more about Dynabeads magnetic beads and KingFisher Sample Purification Systems:
• Dynabeads Protein A beads are also available as a 'ready-to-go' kit with buffers included (Cat. No. 10006D)
• See our immunoprecipitation selection table, data, and references
• Dynabeads magnetic beads selection guide
• See magnets for Dynabeads magnetic beads separations
• KingFisher automation protocols
• Watch a video about the KingFisher Flex instrument

*Sepharose is a trademark of GE Healthcare Bio-Sciences AB.

Specifications

Specifications

ISO9001 and ISO13485
Store at 2–8°C.

Recombinant Protein A covalently bound to Dynabeads
Polystyrene
Beads in suspension
High-throughput Compatible
Protein A
0.02% NaN3
Antibodies
Ambient temperature
Monosized 2.8 mm (CV <5%)
2.8 μm
30 mg/mL
Dynabeads Protein A are supplied in PBS, pH 7.4 w/0.01% Tween-20, 0.09% NaN3
Immunoprecipitation
Superparamagnetic Beads
KingFisher™ Sample Purification System, DynaMag™ magnets
Magnetic Beads
1000 Tests
Dynabeads™
For Research Use Only
Protein A
50 mL
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.