missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ DYNAL™ Dynabeads™ M-280 Sheep Anti-Rabbit IgG, Polyclonal DFS Item

2.8μm superparamagnetic beads with affinity purified polyclonal sheep anti-rabbit IgG covalently bound to bead surface

$614.00 - $1,612.00

Specifications

Storage Requirements Storage: 2°C to 8°C.
View More Specs

Includes: 2mL (10mg/mL concentration) Dynabeads M-280 Sheep anti-Rabbig IgG

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11203D View Documents Invitrogen™
LS11203D
2mL Each for $614.00
11204D View Documents Invitrogen™
LS11204D
10mL Each for $1,612.00
Description

Description

Dynabeads M-280 Sheep anti-Rabbit IgG beads are designed to serve as solid support for simple and efficient binding of rabbit IgG subclasses and their target proteins.

  • Fast and easy coupling and isolation procedure
  • No columns or centrifugation steps required
  • Binds most rabbit IgG subclasses
  • Whole Antibody
  • Capacity: Processes approx. 2 x 109 cells total
  • Cell Type: All cells from all species, except rabbit
  • Concentration: 10mg/ml
  • Isolation Method: Depletion, negative isolation or positive isolation
  • Output Viability: >95%
  • Reactivity: All species, except rabbit

  • Magnetic bead-based separation offers easy handling
  • Adaptable for various applications

Antibody Purification, Bead-Based IVD Assay Development, Bead-Based Immunoassay IVD, Cell Analysis, Cell Isolation, Cell Isolation and Expansion, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Clinical, Diagnostic Development, Human Cell Isolation, Immunoprecipitation, Molecular Diagnostic Test Development, Mouse Cell Isolation, Organelle Isolation, Phage Display, Protein and Peptide Purification, Protein Complex Isolation and Analysis, Protein Complex Pull-Down, Protein Sample Preparation and Protein Purification, Protein-Protein Interactions, Proteins, Expression, Isolation and Analysis, RNAi, Epigenetics and Non-Coding RNA Research, Secondary Cell Isolation

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Storage: 2°C to 8°C.

For Research Use Only. Not for use in diagnostic procedures.