missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Novex™ DYNAL™ Dynabeads™ His-Tag Isolation and Pulldown DFS Item

Designed for his-tagged protein isolation

$309.00 - $1,362.00

Specifications

Concentration 40mg/mL
Format Beads in Suspension
For Use With (Application) Automated Protocols, High Throughput-Compatible
View More Specs

Includes: Protocols. Dynabeads™ His-Tag Isolation and Pulldown beads are supplied as a suspension supplied in 20% ethanol.

Requires: Buffers


Products 2 Description

Description

High selectivity for poly-histidine tags results in extremely low background levels. Bead surface chemistry is cobalt-based and offers exceptionally fast binding kinetics and a high yield per microliter of beads. The entire protocol (from cleared lysate to purified protein) takes only 20 minutes. Protocol includes instructions for making the required buffers and methods for optimizing buffer composition. Dynabeads His-Tag Isolation and Pulldown replaces Dynabeads TALON™. Dynabeads His-Tag Isolation and Pulldown has a optimized binding chemistry that offers significant advantages over Dynabeads TALON which were coated in the TALON chemistry. This optimization results in a cleaner prep with higher yield of his-tag protein and extremely low background levels. Dynabeads His-Tag Isolation and Pulldown is designed for isolating His-tagged proteins from cleared lysates. Isolated his-tagged proteins can either be eluted from the beads or kept on the beads for use in bead-based downstream assays. Bead bound his-tagged proteins can be used in various downstream assays such as bait-prey pulldowns, etc.

Technology used for discontinued Dynabeads TALON was licensed from Clontech. TALON is a registered trademark of Clontech Laboratories Inc., USA.

His-Tagged Protein Purification, Protein and Peptide Purification, Protein Complex Pull-Down, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

40mg/mL
Beads in Suspension
Automated Protocols, High Throughput-Compatible

For Research Use Only. Not for use in diagnostic procedures.