Honeywell™ North™Flexible Low Profile P100 Filters
Thin, high-performance respirator filters are P100 low profile, flexible filters. Available with or without nuisance odor relief.
$9.76 - $757.07
Specifications
Cartridge Type | Filter Cartridge |
---|---|
Certifications/Compliance | NIOSH |
Color | Magenta |
Quantity | 50/Cs. |
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
19-003263
|
Honeywell™
75FFP100 |
Filter |
Pack of 2 Each for $9.76
Case of 50 Pack for $419.50
|
|
|||||
19-161-841
|
Honeywell™
75FFP100NL |
Respirator Filter Plus Odor Relief |
Case of 50 Each for $757.07
|
|
|||||
Description
Cartridges for use with 5500, RU8500 and 7700 Series half masks as well as 5400, RU6500 and 7600 Series full facepieces.
Specifications
Filter Cartridge | |
NIOSH | |
Magenta | |
50/Cs. |