Honeywell™ North™ A600 Series Goggles
Fit over most prescription eyewear
$4.87 - $8.24
Specifications
Certifications/Compliance | ANSI Z87.1-2010 and CSA Z94.3 standards |
---|---|
Lens Tint | Clear |
Frame Color | Transparent Green |
Material | Polycarbonate |
Catalog Number | Mfr. No. | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Price | Quantity | ||||||
29-635-6004
|
Honeywell™
UVA610S |
N/A
|
|
||||||
18-813-324
|
Honeywell™
A610I |
N/A
|
|
||||||
Description
- Transparent green body with clear lens
- Fitted with elastic headband
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
Specifications
ANSI Z87.1-2010 and CSA Z94.3 standards | |
Clear | |
Transparent Green | |
Polycarbonate |