missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitric Acid, ICP-OES, for Trace Metal Analysis, Thermo Scientific

Suitable for digestion of samples prior to trace metal analysis via ICP-OES. Tested for 34 metals at ppb levels and also meets ACS testing specifications

Manufacturer:  Thermo Scientific T003092500

 View more versions of this product

Catalog No. T003092500



Description

Description

  • Suitable for trace metal analysis by ICP-OES
  • Tested for 34 metal impurities at ppb levels
  • Also meets general ACS testing specifications
  • Ideal for analysis of environmental samples
  • Offered in 500mL and 2.5L poly coated glass bottles
Specifications

Specifications

≤ 30 ppb
10 max. (APHA)
7697-37-2,7732-18-5
≤ 5 ppb
≤ 1 ppb
For trace metal analysis
HNO3
≤ 1 ppb
≤ 0.05 ppm
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
63.01
CHEBI:48107
69.0 - 70.0%
≤ 4 ppb
≤ 10 ppb
≤ 1 ppb
≤ 7 ppb
≤ 5 ppb
Poly coated glass bottle
≤ 5 ppb
≤ 1 ppb
≤ 1 ppb
≤ 5 ppb
≤ 5 ppb
Nitric Acid
2.5L
Clear, colorless liquid
≤ 1 ppb
≤ 4 ppb
≤ 1 ppb
≤ 50 ppb
≤ 10 ppb
acide nitrique
[N+](=O)(O)[O-]
1.42
944
≤ 1 ppb
ICP-OES
100 ppb max.
≤ 0.5 ppb
≤ 1 ppb
≤ 0.5 ppb
≤ 1 ppb
≤ 0.10 ppm
≤ 2 ppm
≤ 200 ppb
≤ 0.4 ppm
≤ 1 ppb
≤ 1 ppb

Chemical Identifiers

7697-37-2
63.01
acide nitrique
CHEBI:48107
[N+](=O)(O)[O-]
HNO3
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
SDS Documents