Learn More
Nisin, 900 I.U./mg, Thermo Scientific Chemicals
Nisin, CAS # 1414-45-5, is a polycyclic peptide derived from and produced by Lactococcus lactis that shows antibacterial activity against gram-positive bacteria.
$94.89 - $918.78
Chemical Identifiers
CAS | 1414-45-5 |
---|---|
Molecular Formula | C143H230N42O37S7 |
Molecular Weight (g/mol) | 3354.09 |
MDL Number | MFCD00131724 |
InChI Key | NVNLLIYOARQCIX-OAXMGIFASA-N |
PubChem CID | 131847819 |
SMILES | CCC(C)C1C(=O)NC(=C)C(=O)NC(C(=O)NC(CSCC(C(=O)N1)NC(=O)C(=CC)NC(=O)C(C(C)CC)N)C(=O)NC2C(SCC(NC(=O)CNC(=O)C3CCCN3C2=O)C(=O)NC(CCCCN)C(=O)NC4C(SCC(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC4=O)C)CC(C)C)CCSC)C(=O)NC(CC(=O)N)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC5C( |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6637003
|
Thermo Scientific Chemicals
J6637003 |
1 g |
Each for $94.89
|
|
|||||
AAJ6637006
|
Thermo Scientific Chemicals
J6637006 |
5 g |
Each for $277.78
|
|
|||||
AAJ6637014
|
Thermo Scientific Chemicals
J6637014 |
25 g |
Each for $918.78
|
|
|||||
Description
Nisin, 900 I.U./mg is commonly used as a food preservative. It is also used as a selective agent in cell culture for the isolation of gram-negative bacteria, yeast, and molds. Nisin has been used in bioassays to study posttranslational modification. It binds and interferes with the permeability of the cytoplasmic membrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1414-45-5 | |
3354.09 | |
NVNLLIYOARQCIX-OAXMGIFASA-N | |
CCC(C)C1C(=O)NC(=C)C(=O)NC(C(=O)NC(CSCC(C(=O)N1)NC(=O)C(=CC)NC(=O)C(C(C)CC)N)C(=O)NC2C(SCC(NC(=O)CNC(=O)C3CCCN3C2=O)C(=O)NC(CCCCN)C(=O)NC4C(SCC(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC4=O)C)CC(C)C)CCSC)C(=O)NC(CC(=O)N)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC5C( |
C143H230N42O37S7 | |
MFCD00131724 | |
131847819 |
Specifications
Nisin | |
1414-45-5 | |
MFCD00131724 | |
Soluble in water | |
CCC(C)C1C(=O)NC(=C)C(=O)NC(C(=O)NC(CSCC(C(=O)N1)NC(=O)C(=CC)NC(=O)C(C(C)CC)N)C(=O)NC2C(SCC(NC(=O)CNC(=O)C3CCCN3C2=O)C(=O)NC(CCCCN)C(=O)NC4C(SCC(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC4=O)C)CC(C)C)CCSC)C(=O)NC(CC(=O)N)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC5C( | |
131847819 | |
Brown | |
>110°C (230°F) | |
Nisin, 900 I.U./mg |
Powder | |
C143H230N42O37S7 | |
14,6564 | |
NVNLLIYOARQCIX-OAXMGIFASA-N | |
3354.09 | |
3354.06 | |
1 g | |
2967°C |
Safety and Handling
EINECSNumber : 215-807-5
RTECSNumber : QU3340000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only