missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Nigrosine, pure, water soluble, high purity biological stain
$47.75 - $119.62
Chemical Identifiers
CAS | 8005-03-6 |
---|---|
Molecular Formula | C22H14N6Na2O9S2 |
Molecular Weight (g/mol) | 616.49 |
MDL Number | MFCD00044681 |
InChI Key | HKBVRFLHNUEVRO-HMDKGIQMSA-L |
Synonym | Acid Black 2, C.I. 50420 |
PubChem CID | 44134531 |
IUPAC Name | disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate |
SMILES | C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC189480250
|
Thermo Scientific Chemicals
189480250 |
25 g | Glass bottle |
Each for $47.75
|
|
||||
AC189481000
|
Thermo Scientific Chemicals
189481000 |
100 g | Glass bottle |
Each for $119.62
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
8005-03-6 | |
616.49 | |
HKBVRFLHNUEVRO-HMDKGIQMSA-L | |
44134531 | |
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
C22H14N6Na2O9S2 | |
MFCD00044681 | |
Acid Black 2, C.I. 50420 | |
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate |
Specifications
8005-03-6 | |
MFCD00044681 | |
Acid Black 2, C.I. 50420 | |
HKBVRFLHNUEVRO-HMDKGIQMSA-L | |
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] | |
616.49 | |
Pure | |
Complies | |
25 g | |
Nigrosine, Water soluble, Pure |
C22H14N6Na2O9S2 | |
12, 130 | |
Solubility in water: soluble. | |
Authentic | |
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate | |
44134531 | |
Glass bottle | |
Black | |
Crystalline Powder |
RUO – Research Use Only