Learn More
Nicotinic acid, 99.5%, Thermo Scientific Chemicals
$49.52 - $1917.22
Chemical Identifiers
CAS | 59-67-6 |
---|---|
Molecular Formula | C6H5NO2 |
Molecular Weight (g/mol) | 123.11 |
MDL Number | MFCD00006391 |
InChI Key | PVNIIMVLHYAWGP-UHFFFAOYSA-N |
Synonym | nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil |
PubChem CID | 938 |
ChEBI | CHEBI:15940 |
IUPAC Name | pyridine-3-carboxylic acid |
SMILES | OC(=O)C1=CC=CN=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC128290050
|
Thermo Scientific Chemicals
128290050 |
5 g | Glass Bottle |
Each for $49.52
|
|
||||
AC128291000
|
Thermo Scientific Chemicals
128291000 |
100 g | Plastic bottle |
Each for $53.72
|
|
||||
AC128295000
|
Thermo Scientific Chemicals
128295000 |
500 g | Plastic bottle |
Each for $168.82
|
|
||||
AC128290025
|
Thermo Scientific Chemicals
128290025 |
2.5 kg | Plastic bottle |
Each for $524.39
|
|
||||
AC128290100
|
Thermo Scientific Chemicals
128290100 |
10 kg | Plastic drum |
Each for $1,917.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59-67-6 | |
123.11 | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
938 | |
pyridine-3-carboxylic acid |
C6H5NO2 | |
MFCD00006391 | |
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil | |
CHEBI:15940 | |
OC(=O)C1=CC=CN=C1 |
Specifications
59-67-6 | |
100.0 | |
White | |
Authentic | |
Glass Bottle | |
MFCD00006391 | |
22,38 | |
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
pyridine-3-carboxylic acid | |
938 | |
123.11 | |
Powder |
99.4 | |
236.0°C to 239.0°C | |
1% max. (105°C, 1 hr.) | |
99.5% | |
C6H5NO2 | |
5 g | |
15,661 | |
Solubility in water: 17g/L (20°C). Other solubilities: slightly soluble in ethanol (10g/L at 20°C),eth,er,soluble in propylene glycol,in alkali hydroxides,and carbonates,freely soluble in boiling water or boiling ethanol | |
OC(=O)C1=CC=CN=C1 | |
123.11 | |
CHEBI:15940 | |
99.5% | |
Nicotinic acid, 99.5% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 200-441-0
RTECSNumber : QT0525000
TSCA : TSCA
RUO – Research Use Only