Learn More
Nicotinic acid, 99.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 128295000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nicotinic acid | |
59-67-6 | |
100.0 | |
White | |
Authentic | |
Plastic bottle | |
MFCD00006391 | |
22,38 | |
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
pyridine-3-carboxylic acid | |
938 | |
123.11 | |
Powder |
99.5% | |
99.4 | |
236.0°C to 239.0°C | |
1% max. (105°C, 1 hr.) | |
99.5% | |
C6H5NO2 | |
500 g | |
15,661 | |
Solubility in water: 17g/L (20°C). Other solubilities: slightly soluble in ethanol (10g/L at 20°C),eth,er,soluble in propylene glycol,in alkali hydroxides,and carbonates,freely soluble in boiling water or boiling ethanol | |
OC(=O)C1=CC=CN=C1 | |
123.11 | |
CHEBI:15940 | |
99.5% |
Chemical Identifiers
59-67-6 | |
123.11 | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
938 | |
pyridine-3-carboxylic acid |
C6H5NO2 | |
MFCD00006391 | |
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil | |
CHEBI:15940 | |
OC(=O)C1=CC=CN=C1 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 200-441-0
RTECSNumber : QT0525000
TSCA : TSCA
RUO – Research Use Only