Learn More
Nickel(II) chloride hexahydrate, 98%, Thermo Scientific Chemicals
$77.62 - $867.64
Chemical Identifiers
CAS | 7791-20-0 |
---|---|
Molecular Formula | Cl2Ni·6H2O |
MDL Number | MFCD00149809 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1436630
|
Thermo Scientific Chemicals
A1436630 |
250 g |
Each for $77.62
|
|
|||||
AAA143660B
|
Thermo Scientific Chemicals
A143660B |
1000 g |
Each for $259.12
|
|
|||||
AAA143660I
|
Thermo Scientific Chemicals
A143660I |
5000 g |
Each for $867.64
|
|
|||||
Description
Nickel(II) chloride is used in electroplating and as a catalyst for organic conversions, for example in chemo-selective thioacetalization of aldehydes. In combination with lithium aluminum hydride, it serves as a reducing agent for alkenes, alkynes, and organic halides; it can cleave N-O bond and open epoxides. It is used to prepare a wide array of complexes since water ligands can be easily replaced by amines, thioether, ammonia, and thiolates. It is a precursor to several nickel-phosphine complexes, such as bis(triphenylphosphine)nickel(II) chloride, which are used in alkyne trimerizations, carbonylations, and as catalysts in organic reactions such as Suzuki-Miyaura cross coupling reactions as an alternative to palladium(0) catalysts. It is the precursor to acetylacetonate complex of Ni, used for producing 1,5-cyclooctadiene complex, an important reagent in organonickel chemistry. It can be used to prepare the sandwich compound nickelocene through dimethoxyethane complex of nickel chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNickel(II) chloride is used in electroplating and as a catalyst for organic conversions, for example in chemo-selective thioacetalization of aldehydes. In combination with lithium aluminum hydride, it serves as a reducing agent for alkenes, alkynes, and organic halides; it can cleave N-O bond and open epoxides. It is used to prepare a wide array of complexes since water ligands can be easily replaced by amines, thioether, ammonia, and thiolates. It is a precursor to several nickel-phosphine complexes, such as bis(triphenylphosphine)nickel(II) chloride, which are used in alkyne trimerizations, carbonylations, and as catalysts in organic reactions such as Suzuki-Miyaura cross coupling reactions as an alternative to palladium(0) catalysts. It is the precursor to acetylacetonate complex of Ni, used for producing 1,5-cyclooctadiene complex, an important reagent in organonickel chemistry. It can be used to prepare the sandwich compound nickelocene through dimethoxyethane complex of nickel chloride.
Notes
Store away from incompatible substances such as peroxides, strong acids and potassium.
Chemical Identifiers
7791-20-0 | |
Cl2Ni·6H2O | |
MFCD00149809 |
Specifications
140°C (in 2H2O) | |
4.9 | |
Cl2Ni·6H2O | |
MFCD00149809 | |
14,6505 | |
237.71 (129.60 Anhydrous) | |
Odorless | |
Nickel(II) chloride hexahydrate |
7791-20-0 | |
250 g | |
NiCl2·6H2O | |
UN3288 | |
Freely soluble in water. Soluble in ethylene glycol,ethanol,and ammonium hydroxide. | |
98% | |
1.92 |
Safety and Handling
GHS H Statement
H301-H331-H334-H350-H360-H372-H341-H315-H317
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c
H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 231-743-0
RTECSNumber : QR6480000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only