Learn More
Naphthalene, 99%, Thermo Scientific Chemicals
$45.79 - $321.71
Chemical Identifiers
CAS | 91-20-3 |
---|---|
Molecular Formula | C10H8 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00001742 |
InChI Key | UFWIBTONFRDIAS-UHFFFAOYSA-N |
Synonym | naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene |
PubChem CID | 931 |
ChEBI | CHEBI:16482 |
IUPAC Name | naphthalene |
SMILES | C1=CC2=CC=CC=C2C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC180902500
|
Thermo Scientific Chemicals
180902500 |
250 g | Plastic bottle |
Each for $45.79
|
|
||||
AC180900010
|
Thermo Scientific Chemicals
180900010 |
1 kg | Plastic bottle |
Each for $86.10
|
|
||||
AC180900025
|
Thermo Scientific Chemicals
180900025 |
2.5 kg | Plastic bottle |
Each for $182.00
|
|
||||
AC180900050
|
Thermo Scientific Chemicals
180900050 |
5 kg | Plastic drum |
Each for $321.71
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
91-20-3 | |
128.17 | |
UFWIBTONFRDIAS-UHFFFAOYSA-N | |
931 | |
naphthalene |
C10H8 | |
MFCD00001742 | |
naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene | |
CHEBI:16482 | |
C1=CC2=CC=CC=C2C=C1 |
Specifications
91-20-3 | |
100.0 | |
White | |
218.0°C | |
Authentic | |
Plastic bottle | |
C10H8 | |
250 g | |
0.99 | |
naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene | |
UFWIBTONFRDIAS-UHFFFAOYSA-N | |
naphthalene | |
931 | |
128.17 | |
Crystalline Flakes, Crystals, Tablets, Balls or Crystalline Powder |
98.5 | |
79.0°C to 82.0°C | |
0.9900g/mL | |
78°C | |
98.5% min. (GC) | |
C10H8 | |
MFCD00001742 | |
05,IV,164 | |
15,6455 | |
Solubility in water: 30mg/l (25°C). Other solubilities: soluble in ethanol,,very soluble in acetone,chloroform,ether,,benzene,toluene,carbon disulfide | |
C1=CC2=CC=CC=C2C=C1 | |
128.17 | |
CHEBI:16482 | |
99% | |
Naphthalene |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Flammable solid.
GHS P Statement
Obtain special instructions before use.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smokin
GHS Signal Word: Warning
EINECSNumber : 202-049-5
RUO – Research Use Only