missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6206806
Description
Nalpha, Nepsilon-Di-Boc-L-lysine N-succinimidyl ester is used as a local anesthetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNalpha, Nepsilon-Di-Boc-L-lysine N-succinimidyl ester is used as a local anesthetic.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
184°C | |
30189-36-7 | |
C20H33N3O8 | |
1559007 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O | |
443.50 | |
443.49 | |
5 g | |
Powder |
Nalpha,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester | |
White | |
MFCD00057898 | |
boc-lys boc-osu, 2,5-dioxoazolidinyl 2s-2,6-bis tert-butoxy carbonylamino hexanoate, 2,5-dioxopyrrolidin-1-yl n∼2∼,n∼6∼-bis tert-butoxycarbonyl-l-lysinate, s-2,5-dioxopyrrolidin-1-yl 2,6-bis tert-butoxycarbonyl amino hexanoate, a,n, n, n,n-di-boc-l-lysine hydroxysuccinimide ester, a-di-boc-l-lysine hydroxysuccinimide ester, n,n-di-boc-l-lysine hydroxysuccinimide ester, boc-lys boc-osu calc. based on dry substance, c/n, pubchem14936, boc-lys boc-osu calc. based on dry substance, c/n | |
IQVLXQGNLCPZCL-UGPWUYPHNA-N | |
(2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate | |
13875534 | |
97% | |
Moisture sensitive |
Chemical Identifiers
30189-36-7 | |
443.50 | |
IQVLXQGNLCPZCL-UGPWUYPHNA-N | |
13875534 | |
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O |
C20H33N3O8 | |
MFCD00057898 | |
boc-lys boc-osu, 2,5-dioxoazolidinyl 2s-2,6-bis tert-butoxy carbonylamino hexanoate, 2,5-dioxopyrrolidin-1-yl n∼2∼,n∼6∼-bis tert-butoxycarbonyl-l-lysinate, s-2,5-dioxopyrrolidin-1-yl 2,6-bis tert-butoxycarbonyl amino hexanoate, a,n, n, n,n-di-boc-l-lysine hydroxysuccinimide ester, a-di-boc-l-lysine hydroxysuccinimide ester, n,n-di-boc-l-lysine hydroxysuccinimide ester, boc-lys boc-osu calc. based on dry substance, c/n, pubchem14936, boc-lys boc-osu calc. based on dry substance, c/n | |
(2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only