Learn More
Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Utilize these chemically inert, translucent and autoclavable funnels which feature a TFE stopcock assembly. Chemical-resistant, leakproof and requires no lubrication.
$2337.70 - $4512.12
Catalog Number | Mfr. No. | Description | Capacity (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Capacity (Metric) | Price | Quantity | ||||
10-437-25A
|
Thermo Scientific™
43010125 |
Separatory Funnel FEP 125mL; 28mm closure | 125 mL |
Case of 4 Each for $2,337.70
|
|
||||
10-437-25B
|
Thermo Scientific™
43010250 |
Separatory Funnel FEP 250mL; 33mm closure | 250 mL |
Case of 4 Each for $2,702.27
|
|
||||
10-437-25C
|
Thermo Scientific™
43010500 |
Separatory Funnel FEP 500mL; 43mm closure | 500 mL |
Case of 4 Each for $3,645.94
|
|
||||
10-437-25D
|
Thermo Scientific™
43011000 |
Separatory Funnel FEP 1000mL; 53mm closure | 1000 mL |
Case of 4 Each for $4,512.12
|
|
||||
10-437-25E
|
Thermo Scientific™
43012000 |
Separatory Funnel FEP 2000mL; 53mm | 2000 mL |
Case of 2 Each for $3,262.30
|
|
||||
Description
Perform your liquid-liquid extractions with these chemically inert, translucent and autoclavable Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer and Closure made with Tefzel™ which feature a TFE stopcock assembly. TFE stopcock assembly has excellent chemical resistance, is leakproof† when used with provided Nalgene closure and does not require lubrication — can be chemically disinfected. More transparent than our polypropylene separatory funnels. Compatible with all common lab reagents and solvents. Won't chip, crack or shatter like glass separatory funnels.
Features:- Break-resistant and impervious to any chemical used in a separatory funnel
- No phthalates to contaminate analyses
- Useful in radiological and hazardous separations
- Autoclavable/Leakproof† when used with provided Nalgene closure
- Stem length: 65mm
Ordering Information:
For replacement stopcock assembly for 125mL to 1L sizes, order Cat. No. 714301-1025. For replacement stopcock assembly for 2L size, order Cat. No. 714301-1050.
Note:
Funnel can be autoclaved; stopcock can be chemically sterilized. No stopcock lubrication needed.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
Teflon FEP |
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.