Learn More
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.
$56.42 - $821.12
Specifications
Disposable | Single-use |
---|---|
Filter Type | Membrane Filter |
Material | Polystyrene |
Sterility | Sterile |
No. per Case | 12 |
Alert:
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.
Catalog Number | Mfr. No. | Material (Membrane) | Volume (Metric) | Pore Size | Fits Bottle Neck Size (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material (Membrane) | Volume (Metric) | Pore Size | Fits Bottle Neck Size (Metric) | Price | Quantity | ||
09-741-08
|
Thermo Scientific™
5963320 |
PES | 150 mL | 0.2 μm | 33mm |
Case of 12 Each for $311.03
|
|
||
09-741-09
|
Thermo Scientific™
5964520 |
PES | 150 mL | 0.2 μm | 45mm |
Case of 12 Each for $308.29
|
|
||
09-740-62C
|
Thermo Scientific™
2963345 |
PES | 150 mL | 0.45 μm | 33mm |
Case of 12 Each for $56.42
|
|
||
09-740-62D
|
Thermo Scientific™
2964545 |
PES | 150 mL | 0.45 μm | 45mm |
Case of 12 Each for $339.88
|
|
||
09-741-06
|
Thermo Scientific™
5953320 |
PES | 500 mL | 0.2 μm | 33mm |
Case of 12 Each for $425.59
|
|
||
09-741-07
|
Thermo Scientific™
5954520 |
PES | 500 mL | 0.2 μm | 45mm |
Case of 12 Each for $425.59
|
|
||
09-740-64A
|
Thermo Scientific™
2953345 |
PES | 500 mL | 0.45 μm | 33mm |
Case of 12 Each for $462.41
|
|
||
09-740-64B
|
Thermo Scientific™
2954545 |
PES | 500 mL | 0.45 μm | 45mm |
Case of 12 Each for $461.06
|
|
||
09-741-10
|
Thermo Scientific™
5973320 |
PES | 1000 mL | 0.2 μm | 33mm |
Case of 12 Each for $687.50
|
|
||
09-741-11
|
Thermo Scientific™
5974520 |
PES | 1000 mL | 0.2 μm | 45mm |
Case of 12 Each for $679.32
|
|
||
09-740-22D
|
Thermo Scientific™
2903320 |
SFCA | 150 mL | 0.2 μm | 33mm |
Case of 12 Each for $369.65
|
|
||
09-740-22F
|
Thermo Scientific™
2904520 |
SFCA | 150 mL | 0.2 μm | 45mm |
Case of 12 Each for $369.65
|
|
||
09-740-22E
|
Thermo Scientific™
2903345 |
SFCA | 150 mL | 0.45 μm | 33mm |
Case of 12 Each for $376.74
|
|
||
09-740-22G
|
Thermo Scientific™
2904545 |
SFCA | 150 mL | 0.45 μm | 45mm |
Case of 12 Each for $378.03
|
|
||
09-740-22H
|
Thermo Scientific™
2913320 |
SFCA | 500 mL | 0.2 μm | 33mm |
Case of 12 Each for $518.12
|
|
||
09-740-22K
|
Thermo Scientific™
2914520 |
SFCA | 500 mL | 0.2 μm | 45mm |
Case of 12 Each for $518.12
|
|
||
09-740-22J
|
Thermo Scientific™
2913345 |
SFCA | 500 mL | 0.45 μm | 33mm |
Case of 12 Each for $517.26
|
|
||
09-740-22L
|
Thermo Scientific™
2914545 |
SFCA | 500 mL | 0.45 μm | 45mm |
Case of 12 Each for $518.12
|
|
||
09-740-47A
|
Thermo Scientific™
2923320 |
SFCA | 1000 mL | 0.2 μm | 33mm |
Case of 12 Each for $812.56
|
|
||
09-740-47C
|
Thermo Scientific™
2924520 |
SFCA | 1000 mL | 0.2 μm | 45mm |
Case of 12 Each for $821.12
|
|
||
Description
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.
- Available in PES and SFCA membranes. Devices are color coded according to membrane.
- Devices feature the fastest flow rate and lowest clogging of comparable alternatives
- 0.1um PES available for mycoplasma protection
- 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
- Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
- Available with Fit for Purpose Sterile documentation
Specifications
Single-use | |
Polystyrene | |
12 | |
Bottle Top Filter |
Membrane Filter | |
Sterile | |
12 | |
Sterile |