Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.
$123.97 - $184.26
Catalog Number | Mfr. No. | Diameter (Metric)Top | Capacity (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric)Top | Capacity (Metric) | Price | Quantity | ||||
10-347C
|
Thermo Scientific™
42500035 |
35 mm | 10 mL |
Pack of 12 Each for $143.24
Case of 3 Pack for $429.72
|
|
||||
10-347B
|
Thermo Scientific™
42500055 |
54 mm | 41 mL |
Pack of 12 Each for $155.79
Case of 3 Pack for $467.37
|
|
||||
10-347D
|
Thermo Scientific™
42500065 |
63 mm | 64 mL |
Pack of 12 Each for $184.26
Case of 3 Pack for $552.78
|
|
||||
10-347E
|
Thermo Scientific™
42500075 |
76 mm | 114 mL |
Pack of 6 Each for $130.65
Case of 6 Pack for $783.90
|
|
||||
10-347G
|
Thermo Scientific™
42500100 |
98 mm | 254 mL |
Pack of 4 Each for $123.97
Case of 6 Pack for $743.82
|
|
||||
10-347H
|
Thermo Scientific™
42500160 |
158 mm | 976 mL |
Pack of 2 Each for $150.35
Case of 6 Pack for $902.10
|
|
||||
Description
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.
- Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
- Autoclavable
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
Polypropylene |